Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Picolinamide (2-picolinamide) | 1452-77-3 | MFCD00023483 | 100.0% | 122.13 g/mol | C6H6N2O | 100 G
SDP

Catalog No. 5000212033
Encompass_Preferred

Picolinamide (2-picolinamide) is a research-grade small molecule inhibitor of Poly(ADP-ribose) synthetase (PARP) activity in nuclei from rat pancreatic islet cells. Supplied as a high-purity solid, it is intended for biochemical and cell biology research applications related to PARP-mediated pathways.

  • High purity: 99.97% by HPLC.
  • White to off-white solid appearance.
  • Molecular formula C6H6N2O; molecular weight 122.13 g/mol.
  • Available in solid pack sizes up to 100 G.
  • Storage: stable at room temperature for 3 years; in solvent store at -80°C (2 years) or -20°C (1 year).
  • For research use only; not for human or clinical use.

Catalog No. 50-002-12033 Supplier Medchemexpress LLC Supplier No. HY101020100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Picolinamide (2-picolinamide) is a research-grade small molecule inhibitor of Poly(ADP-ribose) synthetase (PARP) activity in nuclei from rat pancreatic islet cells. Supplied as a high-purity solid, it is intended for biochemical and cell biology research applications related to PARP-mediated pathways.

  • High purity: 99.97% by HPLC.
  • White to off-white solid appearance.
  • Molecular formula C6H6N2O; molecular weight 122.13 g/mol.
  • Available in solid pack sizes up to 100 G.
  • Storage: stable at room temperature for 3 years; in solvent store at -80°C (2 years) or -20°C (1 year).
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.