Learn More
Medchemexpress LLC Picolinic-3,4,5,6-d4 acid | 284487-61-2 | MFCD00145121 | 100.0% | 127.14 | C6HD4NO2 | 10 MG
SDP
Catalog No. 5000281606
Encompass_Preferred
Picolinic acid-d4 is the deuterium-labeled isotopologue of picolinic acid used as a stable-isotope reference standard and tracer in analytical workflows. It is supplied with high isotopic enrichment for use as an internal standard in quantitative methods and is intended for research use only.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High isotopic enrichment (99.99%) for accurate tracing.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Available in small laboratory-scale quantities for standards preparation.
  • Provided with product quality data for analytical use.
Catalog No. 50-002-81606 Supplier Medchemexpress LLC Supplier No. HYW259489S10MG
Picolinic acid-d4 is the deuterium-labeled isotopologue of picolinic acid used as a stable-isotope reference standard and tracer in analytical workflows. It is supplied with high isotopic enrichment for use as an internal standard in quantitative methods and is intended for research use only.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High isotopic enrichment (99.99%) for accurate tracing.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Available in small laboratory-scale quantities for standards preparation.
  • Provided with product quality data for analytical use.