Learn More
Selleck Chemical LLC Picolinic acid
SDP
Catalog No. 5000003725
Encompass_Preferred
Picolinic acid (PCL 016) is a pyridine carboxylate metabolite of tryptophan and acts as an anti-infective and immunomodulator It acts by binding with the zinc associated with zinc finger proteins to alter their structure and inhibit function
Catalog No. 50-000-03725 Supplier Selleck Chemical LLC Supplier No. S624925MG
Picolinic acid (PCL 016) is a pyridine carboxylate metabolite of tryptophan and acts as an anti-infective and immunomodulator It acts by binding with the zinc associated with zinc finger proteins to alter their structure and inhibit function