Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Picolinic acid, chromium salt, MP Biomedicals™

Catalog No. ICN19354201 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19354201 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19354201 Supplier MP Biomedicals, Inc Supplier No. 0219354201

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 14639-25-9
Molecular Formula C18H12CrN3O6
Molecular Weight (g/mol) 418.305
InChI Key CBDQOLKNTOMMTL-UHFFFAOYSA-K
Synonym chromium picolinate, chromium iii pyridine-2-carboxylate, chromium iii picolinate, chromium tripicolinate, picolinic acid chromium iii salt, chromium, tris picolinato, unii-s71t8b8z6p, chromium iii trispicolinate, picolinic acid, chromium salt, chromium 2-pyridinecarboxylate
PubChem CID 56840998
ChEBI CHEBI:50369
IUPAC Name chromium(3+);pyridine-2-carboxylate
SMILES C1=CC=NC(=C1)C(=O)[O-].C1=CC=NC(=C1)C(=O)[O-].C1=CC=NC(=C1)C(=O)[O-].[Cr+3]

Specifications

Color Red
Quantity 1 g
Formula Weight 418.3
Physical Form Powder
Chemical Name or Material Picolinic acid, chromium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.