Learn More
Agilent Technologies PICOMAXX HIGH FIDELITY PCR SY
Catalog No. NC9282043
Encompass
PicoMaxx High Fidelity PCR System, 500U, provides sensitive, efficient amplification on a variety of targets up to 10kb, Contains: 10X Reaction buffer
Catalog No. NC9282043 Supplier Agilent Technologies Supplier No. 600422
PicoMaxx High Fidelity PCR System, 500U, provides sensitive, efficient amplification on a variety of targets up to 10kb, Contains: 10X Reaction buffer