Learn More
World Precision Instrument PicoNozzle Gasket, 1.0mm, Green, Pkg of 10-75122-110
SDP
Catalog No. 502521229
Encompass
Package of 10 single size gaskets for use with the PV820, PV830, MMP, and DMP.
Catalog No. 50-252-1229 Supplier World Precision Instrument Supplier No. 75122110
Package of 10 single size gaskets for use with the PV820, PV830, MMP, and DMP.