Learn More
World Precision Instrument PicoNozzle Gasket, 1.2mm, Black, Pkg of 10-75122-210
SDP
Catalog No. 502521228
Encompass
Package of 10 single size gaskets for use with the PV820, PV830, MMP, and DMP.
Catalog No. 50-252-1228 Supplier World Precision Instrument Supplier No. 75122210
Package of 10 single size gaskets for use with the PV820, PV830, MMP, and DMP.