Learn More
World Precision Instrument PicoNozzle Gasket, 1.65mm, White,Pkg of 10-75122-410
SDP
Catalog No. 502521226
Encompass
Package of 10 single size gaskets for use with the PV820, PV830, MMP, and DMP.
Catalog No. 50-252-1226 Supplier World Precision Instrument Supplier No. 75122410
Package of 10 single size gaskets for use with the PV820, PV830, MMP, and DMP.