Learn More
World Precision Instrument PicoNozzle Kit v1-5430-10
SDP
Catalog No. 502521258
Encompass
The PicoNozzle Kit (version 1) allows micropipettes to be securely mounted in micropositioners for stable axial air delivery. Because air enters the pipette axially, lateral whipping during injection is eliminated.
Catalog No. 50-252-1258 Supplier World Precision Instrument Supplier No. 543010
The PicoNozzle Kit (version 1) allows micropipettes to be securely mounted in micropositioners for stable axial air delivery. Because air enters the pipette axially, lateral whipping during injection is eliminated.