Learn More
World Precision Instrument PicoNozzle Kit v1-5430-15
SDP
Catalog No. 502521257
Encompass
The PicoNozzle Kit (version 1) allows micropipettes to be securely mounted in micropositioners for stable axial air delivery. Because air enters the pipette axially, lateral whipping during injection is eliminated.
Catalog No. 50-252-1257 Supplier World Precision Instrument Supplier No. 543015
The PicoNozzle Kit (version 1) allows micropipettes to be securely mounted in micropositioners for stable axial air delivery. Because air enters the pipette axially, lateral whipping during injection is eliminated.