Learn More
World Precision Instrument PicoNozzle Kit v2
SDP
Catalog No. 502521255
Encompass
The PicoNozzle Kit (version 2), the 5430-ALL, allows micropipettes to be securely mounted in micropositioners for stable axial air delivery. Because air enters the pipette axially, lateral whipping during injection is eliminated.
NOTE: Each PicoPump is supplied with a 5430-ALL kit.
Catalog No. 50-252-1255 Supplier World Precision Instrument Supplier No. 5430ALL
The PicoNozzle Kit (version 2), the 5430-ALL, allows micropipettes to be securely mounted in micropositioners for stable axial air delivery. Because air enters the pipette axially, lateral whipping during injection is eliminated.
NOTE: Each PicoPump is supplied with a 5430-ALL kit.