Learn More
Medchemexpress LLC Picoplatin | 181630-15-9 | MFCD31735135 | 98.5% | C6H10Cl2N2Pt | 10MG
SDP
Supplier:  Medchemexpress LLC HY13146810MG
Encompass_Preferred
Picoplatin (AMD473) is a platinum-based antineoplastic agent used in research to form cytotoxic DNA lesions while reducing thiol-mediated drug resistance. Supplied as a solid with documented purity and storage recommendations, it is intended for laboratory studies of platinum drug activity and resistance mechanisms.
  • Platinum-based antineoplastic research compound.
  • Designed to reduce thiol-mediated drug resistance.
  • Forms cytotoxic lesions with DNA.
  • High chemical purity (98.5%).
  • Supplied in small milligram quantities for research use.
  • Solid, light yellow to yellow appearance.
  • Recommended storage sealed and away from moisture.
Catalog No. 50-002-58893