Learn More
Cayman Chemical Picosulfate sodium salt hydr
SDP
Catalog No. 502948187
Encompass_Preferred
A stimulant laxative and prodrug form of desacetyl bisacodyl; increases excreted fecal weight in a rat model of loperamide-induced constipation
Catalog No. 50-294-8187 Supplier Cayman Chemical Supplier No. 34566250 MG
A stimulant laxative and prodrug form of desacetyl bisacodyl; increases excreted fecal weight in a rat model of loperamide-induced constipation