Learn More
Cayman Chemical Picosulfate sodium salt hydr
SDP
Supplier:  Cayman Chemical 34566100 MG
Encompass_Preferred
A stimulant laxative and prodrug form of desacetyl bisacodyl; increases excreted fecal weight in a rat model of loperamide-induced constipation
Catalog No. 50-294-8186