Learn More
Waters Corp Chemistry Package, Amino Acid Analysis, HPLC, UV Detection
Supplier:  Waters Corp WAT007360
Encompass_Preferred
This all-inclusive PicoTag Chemistry Package supports the widely used PicoTag Method for amino acid analysis, applicable to protein hydrolysates, physiological fluids, feeds, foods, and pharmaceutical preparations.
  • Supports 250 analyses.
  • Utilizes HPLC separation mode and UV detection.
  • Includes column reagent kit, eluents, diluents, manual, column heater inserts, and sample tubes.
  • Reagent kit contains PITC, TEA, and standards A/N and B.
  • Provided with Eluent 1 (4 × 1 L), Eluent 2 (4 × 1 L), and Diluent (100 mL).
  • Storage condition: Room temperature.
Catalog No. 50-784-966