Learn More
Waters Corp Waters Chemistry Package, Amino Acid Analysis, HPLC, 250 Analyses
Supplier:  Waters Corp WAT091681
Encompass_Preferred
The Pico-Tag Method has been widely used for amino acid analysis for over two decades. This all-inclusive chemistry package is designed to streamline the amino acid analysis workflow, providing essential tools, consumables, and materials for successful amino acid quantitation.
  • Streamlines amino acid analysis workflow with specially formulated and QC-processed products.
  • Designed for high-performance liquid chromatography (HPLC) amino acid analysis.
  • Suitable for diverse samples including protein hydrolysates, physiologic fluids, and pharmaceutical preparations.
  • Utilizes pre-column derivatization with phenylisothiocyanate to form stable phenylthiocarbamyl derivatives.
  • Enables easy separation of derivatives by reversed-phase HPLC.
  • Volatile reaction by-products and reagents can be removed by vacuum drying.
Catalog No. 50-785-975
Add to Cart
Request a Quote