Learn More
Waters Corp Waters Corporation Pico-Tag Eluent, Reversed Phase, 4 x 1 L, Amino Acid Application
Supplier: Waters Corp WAT010965
The Pico Tag™ Method has been widely used for amino acid analysis for over two decades. Eluent 2, specifically designed for use in this method, can be ordered separately. This product is supplied in a 4 x 1 L format.
- Separation mode: Reversed phase
- Detection method: Optical
- Formulation: Liquid
- System type: LC
- Shelf life upon reconstitution: 1 month
- Application: Amino acid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.