Learn More
Enzo Life Sciences Picotamide (10 mg). CAS: 32828-81-2
SDP
Supplier:  Enzo Life Sciences BMLRA1040010
Encompass_Preferred
A dual activity thromboxane A2 antagonist and TXA2 synthase inhibitor. Clinically useful antiplatelet agent. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25mg/ml) or ethanol (25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1706