Learn More
Medchemexpress LLC Picotamide | 32828-81-2 | MFCD00866792 | 99.8% | 376.41 g/mol | C21H20N4O3 | 10 MG
SDP
Catalog No. 5000296480
Encompass_Preferred
Picotamide is a dual thromboxane A2 (TxA2) synthase and receptor inhibitor used in preclinical research to study platelet aggregation and cardiovascular disease mechanisms.
  • Combined thromboxane A2 synthase and receptor inhibitor.
  • Exhibits antiplatelet activity for studies of platelet aggregation.
  • Reported to reduce microalbuminuria and inhibit carotid plaque growth in diabetic models.
  • High purity (99.76%) suitable for biochemical assays.
  • Molecular formula C21H20N4O3; molecular weight 376.41 g/mol.
  • Offered in small mg quantities for laboratory use.
Catalog No. 50-002-96480 Supplier Medchemexpress LLC Supplier No. HYB173510MG
Picotamide is a dual thromboxane A2 (TxA2) synthase and receptor inhibitor used in preclinical research to study platelet aggregation and cardiovascular disease mechanisms.
  • Combined thromboxane A2 synthase and receptor inhibitor.
  • Exhibits antiplatelet activity for studies of platelet aggregation.
  • Reported to reduce microalbuminuria and inhibit carotid plaque growth in diabetic models.
  • High purity (99.76%) suitable for biochemical assays.
  • Molecular formula C21H20N4O3; molecular weight 376.41 g/mol.
  • Offered in small mg quantities for laboratory use.