Learn More
Medchemexpress LLC Picoxystrobin | 117428-22-5 | 99.8% | 367.32 | 100 MG
SDP
Catalog No. 5000348442
Encompass_Preferred
Picoxystrobin is a strobilurin fungicide that controls plant diseases by inhibiting mitochondrial respiration. It has been observed to be highly toxic to zebrafish embryos, causing developmental abnormalities, oxidative stress, and immunotoxicity.
  • Controls plant diseases by inhibiting mitochondrial respiration
  • Causes developmental abnormalities in zebrafish embryos
  • Induces oxidative stress in zebrafish embryos
  • Leads to immunotoxicity in zebrafish embryos
Catalog No. 50-003-48442 Supplier Medchemexpress LLC Supplier No. HY136355100MG
Picoxystrobin is a strobilurin fungicide that controls plant diseases by inhibiting mitochondrial respiration. It has been observed to be highly toxic to zebrafish embryos, causing developmental abnormalities, oxidative stress, and immunotoxicity.
  • Controls plant diseases by inhibiting mitochondrial respiration
  • Causes developmental abnormalities in zebrafish embryos
  • Induces oxidative stress in zebrafish embryos
  • Leads to immunotoxicity in zebrafish embryos