Learn More
Medchemexpress LLC Picoxystrobin | 117428-22-5 | 99.8% | 367.32 | 250 MG
SDP
Catalog No. 5000350145
Encompass_Preferred
Picoxystrobin is a strobilurin fungicide that controls plant diseases by inhibiting mitochondrial respiration. It has been shown to be highly toxic to zebrafish embryos, causing developmental abnormalities, oxidative stress, and immunotoxicity. This product is for research use only.
  • Controls plant diseases by inhibiting mitochondrial respiration
  • Can cause developmental abnormalities, oxidative stress, and immunotoxicity in zebrafish embryos
  • For research use only
Catalog No. 50-003-50145 Supplier Medchemexpress LLC Supplier No. 250MGHY136355
Add to Cart
Add to Cart
Picoxystrobin is a strobilurin fungicide that controls plant diseases by inhibiting mitochondrial respiration. It has been shown to be highly toxic to zebrafish embryos, causing developmental abnormalities, oxidative stress, and immunotoxicity. This product is for research use only.
  • Controls plant diseases by inhibiting mitochondrial respiration
  • Can cause developmental abnormalities, oxidative stress, and immunotoxicity in zebrafish embryos
  • For research use only