Learn More
Cayman Chemical Picoxystrobin, Purity: ≥98%, CAS: 117428-22-5, 50 mg
SDP
Catalog No. 502917537
Encompass_Preferred
A strobilurin fungicide; active against the plant pathogenic fungus R. herpotrichoides when used at 0.75 mg/L; induces tail deformities, pericardial edema, and spine curvature in zebrafish embryos at 100 µg/L; toxic to zebrafish embryos (LC50 = 213.8 µg/L)
Catalog No. 50-291-7537 Supplier Cayman Chemical Supplier No. 3954550 MG
A strobilurin fungicide; active against the plant pathogenic fungus R. herpotrichoides when used at 0.75 mg/L; induces tail deformities, pericardial edema, and spine curvature in zebrafish embryos at 100 µg/L; toxic to zebrafish embryos (LC50 = 213.8 µg/L)