Learn More
Newcomer Supply Picric Acid - Acetone 0.1%, 500 ml
SDP
Catalog No. NC9898086
Encompass
Use: Gram Stains, eg. Brown Brenn. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.
Catalog No. NC9898086 Supplier Newcomer Supply Supplier No. 1335A
Use: Gram Stains, eg. Brown Brenn. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.