Learn More
Electron Microscopy Sciences Picric Acid-Acetone Solution 0.1% 500 ML
SDP
Catalog No. 5030026
Encompass_Preferred
Used in Taylor's Method for Gram Positive and Gram Negative Bacteria. See AFIP Manual of Histological Staining Methods, 3rd ed., Ed. L. Luna: New York: McGraw Hill Publications, c. 1968, p.226 or Clark, G.: Staining Procedures, Williams and Wilkins Company, Baltimore, 3rd Ed, c.1973, p.320, or Taylor, R.D., Amer. J. Clin. Path. 46:472 (1966)
Catalog No. 50-300-26 Supplier Electron Microscopy Sciences Supplier No. 2610809
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Used in Taylor's Method for Gram Positive and Gram Negative Bacteria. See AFIP Manual of Histological Staining Methods, 3rd ed., Ed. L. Luna: New York: McGraw Hill Publications, c. 1968, p.226 or Clark, G.: Staining Procedures, Williams and Wilkins Company, Baltimore, 3rd Ed, c.1973, p.320, or Taylor, R.D., Amer. J. Clin. Path. 46:472 (1966)