Learn More
Electron Microscopy Sciences Picric Acid Orange G Solution 500 ML
SDP
Catalog No. 5031793
Encompass_Preferred
Reference: For the staining of fibrin and platelets. Carstairs (1965).
Catalog No. 50-317-93 Supplier Electron Microscopy Sciences Supplier No. 2638103
Reference: For the staining of fibrin and platelets. Carstairs (1965).