Learn More
Electron Microscopy Sciences Picric Acid, Reagent, Crystal, A.C.S. 100 GR
SDP
Catalog No. 5027866
Encompass_Preferred
(2,4,6-Trinitrophenol)
C₆H₂(NO₂) ₃OH F.W. 229.11
Melting point: 121-123°C
Catalog No. 50-278-66 Supplier Electron Microscopy Sciences Supplier No. 19550
(2,4,6-Trinitrophenol)
C₆H₂(NO₂) ₃OH F.W. 229.11
Melting point: 121-123°C