Learn More
Newcomer Supply Picric Acid, Saturated Alcoholic, 500 ml
SDP
Catalog No. NC1413419
Encompass
Use: Differentiator for Sulfated Alcian Blue stain and for formalin pigment removal. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.
Catalog No. NC1413419 Supplier Newcomer Supply Supplier No. 1337A
Use: Differentiator for Sulfated Alcian Blue stain and for formalin pigment removal. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.