Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Picric Acid, Saturated, LabChem™

Catalog No. LC186701 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC186701 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. LC186701 Supplier LabChem, Inc. Supplier No. LC186701

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 88-89-1 , 7732-18-5
Molecular Formula C6H3N3O7
Molecular Weight (g/mol) 229.104
InChI Key OXNIZHLAWKMVMX-UHFFFAOYSA-N
Synonym picric acid, trinitrophenol, carbazotic acid, picronitric acid, melinite, acide picrique, phenol trinitrate, picral, nitroxanthic acid, pikrinsaeure
PubChem CID 6954
ChEBI CHEBI:46149
IUPAC Name 2,4,6-trinitrophenol
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])O)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Color Yellow
CAS Max % 98.7, 1.3
Linear Formula C6H3N3O7
Solubility Information Soluble in water
Formula Weight 229.1
Density 1g/mL
Grade Certified
Quantity 500 mL
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1g/mL
Chemical Name or Material Picric Acid, Saturated
Show More Show Less
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.