Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Picric Acid Solution, Sigma-Aldrich

Catalog No. 01258819 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-819 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-819 Supplier MilliporeSigma Supelco Supplier No. P67441GA
Add to Cart
Add to Cart

Picric acid is commonly used as a fixative and staining reagent in forensic and histological applications. It is a general cytoplasmic stain, connective tissue fiber stain, and a fixative for tissues. It is a component of picrosirius red which is used to stain collagen. Bouin's fluid which also comprises picric acid is a fixative for nuclei and connective tissue. It can be used as a colorimetric reagent in creatinine determination assay.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1.3% in Water
Product Type Fixative
Quantity 1 gal.
Solution Type Fixative
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.