Learn More
Newcomer Supply Picric Acid - Acetone 0.05%, 500 ml
SDP
Catalog No. NC2014187
Encompass
Use: Brown & Hopps. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.
Catalog No. NC2014187 Supplier Newcomer Supply Supplier No. 13351B
Use: Brown & Hopps. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.