Learn More
Abcam Picro-Sirius Red Stain Kit (Cardiac Muscle)
Catalog No. NC1945749
Encompass_Preferred
Picro-Sirius Red Stain Kit (Cardiac Muscle) ab245887 is intended for use in the histological visualization of thin septa and collagen fibers. This modification of the Picro-Sirius Red stain eliminates the yellow cytoplasmic staining that can obscure thin collagenous septa. Using this procedure easily allows viewing of collagenous septa as thin as 0.2-0.5 microns. The PSR stain may be viewed using standard light microscopy or polarized light resulting in birefringence of the collagen fibers.
Catalog No. NC1945749 Supplier Abcam Supplier No. AB2458871KIT
Picro-Sirius Red Stain Kit (Cardiac Muscle) ab245887 is intended for use in the histological visualization of thin septa and collagen fibers. This modification of the Picro-Sirius Red stain eliminates the yellow cytoplasmic staining that can obscure thin collagenous septa. Using this procedure easily allows viewing of collagenous septa as thin as 0.2-0.5 microns. The PSR stain may be viewed using standard light microscopy or polarized light resulting in birefringence of the collagen fibers.