Learn More
Abcam Picro-Sirius Red Solution, 500ML
Catalog No. NC1873312
Encompass_Preferred
Picro-sirius red solution is used for the histological visualization of muscle and collagen fibers in tissue sections, providing clear differentiation under light and polarized light microscopy. - Collagen and muscle fiber staining – collagen stains red, muscle fibers yellow, and cytoplasm yellow under light microscopy - Birefringence – collagen fibers show yellow-orange and green birefringence under polarized light microscopy - Control tissues – suitable for lung, uterus, muscle, and kidney tissue - Form – liquid for easy application
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1873312 Supplier Abcam Supplier No. AB246832500ML
Picro-sirius red solution is used for the histological visualization of muscle and collagen fibers in tissue sections, providing clear differentiation under light and polarized light microscopy. - Collagen and muscle fiber staining – collagen stains red, muscle fibers yellow, and cytoplasm yellow under light microscopy - Birefringence – collagen fibers show yellow-orange and green birefringence under polarized light microscopy - Control tissues – suitable for lung, uterus, muscle, and kidney tissue - Form – liquid for easy application
The product is subject to the following: Abcam Restricted Use Statement