Learn More
Diagnostic Biosystems PICRO SIRIUS RED STAIN KIT Picro Sirius Red Stain Kit
Catalog No. 502407438
Encompass_Preferred
The Picro-Sirius Red Stain Kit (For Collagen) is intended for use in the histological visualization of collagen I and III fibers in addition to muscle in tissue sections.
Catalog No. 50-240-7438 Supplier Diagnostic Biosystems Supplier No. KT037
The Picro-Sirius Red Stain Kit (For Collagen) is intended for use in the histological visualization of collagen I and III fibers in addition to muscle in tissue sections.