Learn More
Cayman Chemical Picrocrocin, Purity: ≥95%, CAS: 138-55-6, 100 mg
SDP
Catalog No. 502978595
Encompass_Preferred
A monoterpene glycoside with anticancer activity; a biosynthetic precursor of safranal; inhibits the growth of HeLa cells (IC50 = 3 mM)
Catalog No. 50-297-8595 Supplier Cayman Chemical Supplier No. 39914100 MG
A monoterpene glycoside with anticancer activity; a biosynthetic precursor of safranal; inhibits the growth of HeLa cells (IC50 = 3 mM)