Learn More
Cayman Chemical PIcrocrocIn 10mg
SDP
Catalog No. 502978655
Encompass_Preferred
A monoterpene glycoside with anticancer activity; a biosynthetic precursor of safranal; inhibits the growth of HeLa cells (IC50 = 3 mM)
Catalog No. 50-297-8655 Supplier Cayman Chemical Supplier No. 3991410 MG
A monoterpene glycoside with anticancer activity; a biosynthetic precursor of safranal; inhibits the growth of HeLa cells (IC50 = 3 mM)