Learn More
Selleck Chemical LLC Picropodophyllin (PPP) 25mg 477-47-4 AXL1717
SDP
Catalog No. NC0929624
Encompass_Preferred
Picropodophyllin (PPP, AXL1717) is a IGF-1R inhibitor with IC50 of 1 nM. It displays selectivity for IGF-1R and does not coinhibit tyrosine phosphorylation the IR, or of a selected panel of receptors less related to IGF-IR(FGF-R, PDGF-R, OR EGF-R). Picropodophyllin (PPP) induces apoptosis with antineoplastic activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0929624 Supplier Selleck Chemical LLC Supplier No. S766825MG
Add to Cart
Add to Cart
Picropodophyllin (PPP, AXL1717) is a IGF-1R inhibitor with IC50 of 1 nM. It displays selectivity for IGF-1R and does not coinhibit tyrosine phosphorylation the IR, or of a selected panel of receptors less related to IGF-IR(FGF-R, PDGF-R, OR EGF-R). Picropodophyllin (PPP) induces apoptosis with antineoplastic activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.