Learn More
Abcam Picropodophylloto x in, IGF-1R inhibitor, 5MG
Catalog No. 502705552
Encompass_Preferred
MW 414.4 Da, Purity >97%. Selective insulin-like growth factor-1 receptor (IGF-1R) inhibitor. Reduces pAkt and pErk1/2. Does not compete with ATP and may inhibit IGF-1R autophosphorylation. Anticancer agent. Active in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5552 Supplier Abcam Supplier No. AB1446235MG
MW 414.4 Da, Purity >97%. Selective insulin-like growth factor-1 receptor (IGF-1R) inhibitor. Reduces pAkt and pErk1/2. Does not compete with ATP and may inhibit IGF-1R autophosphorylation. Anticancer agent. Active in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement