Learn More
Cayman Chemical Picropodophyllotoxin, Purity: ≥98%, CAS: 477-47-4, 10 mg
SDP
Catalog No. 502967024
Encompass_Preferred
A potent and selective inhibitor of IGF-1R (IC50 = 6 nM) causes cell cycle arrest and induces apoptosis in cancer cells both in vitro and in vivo
Catalog No. 50-296-7024 Supplier Cayman Chemical Supplier No. 1732910 MG
A potent and selective inhibitor of IGF-1R (IC50 = 6 nM) causes cell cycle arrest and induces apoptosis in cancer cells both in vitro and in vivo