Learn More
Cayman Chemical PIcrosIde I 10mg
SDP
Catalog No. 502969156
Encompass_Preferred
A natural iridoid glycoside that blocks changes in acid phosphatase activity, phospholipid levels, and lipid peroxide production induced by D-galactosamine in rat liver; also enhances neurite outgrowth in PC12D cells induced by bFGF and 7S NGF when given at 60 μM
Catalog No. 50-296-9156 Supplier Cayman Chemical Supplier No. 1543110 MG
A natural iridoid glycoside that blocks changes in acid phosphatase activity, phospholipid levels, and lipid peroxide production induced by D-galactosamine in rat liver; also enhances neurite outgrowth in PC12D cells induced by bFGF and 7S NGF when given at 60 μM