Learn More
Cayman Chemical PIcrosIde I 25mg
SDP
Catalog No. 502971364
Encompass_Preferred
A natural iridoid glycoside that blocks changes in acid phosphatase activity, phospholipid levels, and lipid peroxide production induced by D-galactosamine in rat liver; also enhances neurite outgrowth in PC12D cells induced by bFGF and 7S NGF when given at 60 μM
Catalog No. 50-297-1364 Supplier Cayman Chemical Supplier No. 1543125 MG
A natural iridoid glycoside that blocks changes in acid phosphatase activity, phospholipid levels, and lipid peroxide production induced by D-galactosamine in rat liver; also enhances neurite outgrowth in PC12D cells induced by bFGF and 7S NGF when given at 60 μM