Learn More
Medchemexpress LLC HY-N0408 5mg Medchemexpress, Picroside II CAS:39012-20-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN04085MG
Encompass_Preferred
Medchemexpress, HY-N0408 5mg Picroside II CAS:39012-20-9 Picroside II, an iridoid compound extracted from Picrorhiza, exhibits anti-inflammatory and anti-apoptotic activities. picroside II alleviates the inflammatory response in sepsis and enhances immune function by inhibiting the activation of NLRP3 inflammasome and NF-κB pathways. Picroside II is an antioxidant, exhibits a significant neuroprotective effect through reducing ROS production and protects the blood-brain barrier (BBB) after cerebral ischemia-reperfusion (CI/R) injury. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0268