Learn More
Polysciences, Inc. Picrosirius Red Stain Kit, 500 mL
SDP
Catalog No. NC9039835
Encompass_Preferred
Kit contains:Solution A - Phosphomolybdic Acid (500mL), 12026-57-2, MFCD00011341 Solution B - Picrosirius Red F3BA Stain (250ml or 500ml) Solution C - 0.1 N Hydrochloride Acid (500mL0, 7647-01-0, MFCD00011324 Picrosirius Red Stain binds specifically to collagen fibrils of varying diameter that is used to distinguish collagen Type I from Type III. Picrosirius Red Stain will quantify the amount of collagen in a given area of myocardial tissue. Picrosirius Red staining is also useful in the quantitative aspect of liver fibrosis by measuring a change in collagen proportion stained as red. The 250 ml kit stains up to 100 slides; 500 ml kit stains up to 200 slides.
Catalog No. NC9039835 Supplier Polysciences, Inc. Supplier No. 24901500
Kit contains:Solution A - Phosphomolybdic Acid (500mL), 12026-57-2, MFCD00011341 Solution B - Picrosirius Red F3BA Stain (250ml or 500ml) Solution C - 0.1 N Hydrochloride Acid (500mL0, 7647-01-0, MFCD00011324 Picrosirius Red Stain binds specifically to collagen fibrils of varying diameter that is used to distinguish collagen Type I from Type III. Picrosirius Red Stain will quantify the amount of collagen in a given area of myocardial tissue. Picrosirius Red staining is also useful in the quantitative aspect of liver fibrosis by measuring a change in collagen proportion stained as red. The 250 ml kit stains up to 100 slides; 500 ml kit stains up to 200 slides.