Learn More
Polysciences, Inc. Picrosirius Red Stain Kit, 250 mL
SDP
Catalog No. NC9908782
Encompass_Preferred
Picrosirius Red Stain binds specifically to collagen fibrils of varying diameter that is used to distinguish collagen Type I from Type III. Picrosirius Red Stain will quantify the amount of collagen in a given area of myocardial tissue. Picrosirius Red staining is also useful in the quantitative aspect of liver fibrosis by measuring a change in collagen proportion stained as red.
Catalog No. NC9908782 Supplier Polysciences, Inc. Supplier No. 24901250
Picrosirius Red Stain binds specifically to collagen fibrils of varying diameter that is used to distinguish collagen Type I from Type III. Picrosirius Red Stain will quantify the amount of collagen in a given area of myocardial tissue. Picrosirius Red staining is also useful in the quantitative aspect of liver fibrosis by measuring a change in collagen proportion stained as red.