Learn More
Medchemexpress LLC HY-107782 1mg Medchemexpress, Picrotin CAS:21416-53-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1077821MG
Encompass_Preferred
Medchemexpress, HY-107782 1mg Picrotin CAS:21416-53-5 Picrotin is an inhibitor of glycine receptors (GlyRs) which blocks α2 GlyR, α1 GlyR and α3 GlyR. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8468