Learn More
Selleck Chemical LLC Picrotoxin-25mg
SDP
Supplier:  Selleck Chemical LLC S610025MG
Encompass_Preferred
Picrotoxin (Picrotin, Cocculin, Cocculus, NSC 403139), a plant alkaloid, is a non-competitive antagonist of neuronal GABA and glycine receptors.
Catalog No. 50-239-0048