Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Picrylsulfonic Acid, 5% (w/v) Aqueous Solution, Spectrum™ Chemical
SDP

Catalog No. 18605218 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
10 mL
5 x 10 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-218 5 x 10 mL Amber Glass Bottle
18-605-217 10 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-605-218 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P328010MLCS5

P3280, 2508-19-2, C6H3N3O9S

Spectrum™ Chemical Picrylsulfonic Acid, 5 Percent (w/v) Aqueous Solution - Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability.

Chemical Identifiers

CAS 2508-19-2 , 7732-18-5
Molecular Formula C6H3N3O9S
Molecular Weight (g/mol) 293.16
InChI Key NHJVRSWLHSJWIN-UHFFFAOYSA-N
IUPAC Name 2,4,6-trinitrobenzene-1-sulfonic acid
SMILES OS(=O)(=O)C1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O

Specifications

CAS Max % 0.05, 0.95
Packaging Amber Glass Bottle
Quantity 5 x 10 mL
UN Number UN3265
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.