Learn More
Medchemexpress LLC HY-50094 200mg , Pictilisib CAS:957054-30-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY50094200MG
Encompass_Preferred
HY-50094 200mg Pictilisib CAS: 957054-30-7 Pictilisib (GDC-0941) is a potent inhibitor of PI3Kα/δ with an IC50 of 3 nM, with modest selectivity against p110β (11-fold) and p110γ (25-fold). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0246