Learn More
Medchemexpress LLC Pictilisib (dimethanesulfonate) | 957054-33-0 | 99.7% | 705.85 | 200 MG
SDP
Catalog No. 5000400705
Encompass_Preferred
Pictilisib dimethanesulfonate is a potent inhibitor of PI3Kα/δ with IC50 of 3 nM, with modest selectivity against p110β (11-fold) and p110γ (25-fold).
  • Potent inhibitor of PI3Kα/δ with IC50 of 3 nM
  • Modest selectivity against p110β (11-fold) and p110γ (25-fold)
  • Reduces tumor cell viability in breast cancer cell lines
  • Inhibits Akt phosphorylation and downstream targets of Akt signaling
  • Shows high efficacy of antitumor activity in ZD1839-resistant non-small cell lung cancer (NSCLC) cell lines (A549 and H460)
  • Highly efficacious in combination with U0126 in inducing cell growth inhibition, G0-G1 arrest, and cell apoptosis
  • Reduces secreted VEGF after hypoxic/anoxic exposure
  • Leads to tumor stasis in animal models
  • Results in tumor regressions during treatment leading to enhanced antitumor responses
  • Reduces tumor growth and PI3K and HIF-1 pathway activity in tumor-bearing mice
Catalog No. 50-004-00705 Supplier Medchemexpress LLC Supplier No. HY20180200MG
Pictilisib dimethanesulfonate is a potent inhibitor of PI3Kα/δ with IC50 of 3 nM, with modest selectivity against p110β (11-fold) and p110γ (25-fold).
  • Potent inhibitor of PI3Kα/δ with IC50 of 3 nM
  • Modest selectivity against p110β (11-fold) and p110γ (25-fold)
  • Reduces tumor cell viability in breast cancer cell lines
  • Inhibits Akt phosphorylation and downstream targets of Akt signaling
  • Shows high efficacy of antitumor activity in ZD1839-resistant non-small cell lung cancer (NSCLC) cell lines (A549 and H460)
  • Highly efficacious in combination with U0126 in inducing cell growth inhibition, G0-G1 arrest, and cell apoptosis
  • Reduces secreted VEGF after hypoxic/anoxic exposure
  • Leads to tumor stasis in animal models
  • Results in tumor regressions during treatment leading to enhanced antitumor responses
  • Reduces tumor growth and PI3K and HIF-1 pathway activity in tumor-bearing mice