Learn More
Perkin Elmer US LLC PID Lamp, 10.2 eV
Catalog No. 502112819
Encompass_Preferred
For most applications including aromatics, alkenes, and aliphatics higher than C4. Shipped standard with PID.
Catalog No. 50-211-2819 Supplier Perkin Elmer US LLC Supplier No. 03303599
For most applications including aromatics, alkenes, and aliphatics higher than C4. Shipped standard with PID.