Learn More
Enzo Life Sciences PIDD monoclonal antibody (Anto-1) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804837C100
Encompass_Preferred
Clone: Anto-1. Isotype: IgG1. Immunogen: Recombinant human PIDD death domain (aa 776-910). Source: Purified from ascites. Species reactivity: Human, Mouse. Applications: IHC (PS), WB. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-9993